Learn More
Abcam Native Human IgM protein, 1MG
Supplier:  Abcam AB2058071MG
Encompass_Preferred
Native Human IgM protein is a Human Full Length protein, in the 1 to 452 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant mu, Ig mu chain C region, Ig mu chain C region BOT, Ig mu chain C region GAL, Ig mu chain C region OU, IGHM
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-8864