Learn More
Abcam Native human Thrombin protein (Active), 100UG
Supplier:  Abcam AB62452100UG
Encompass_Preferred
Native human Thrombin protein (Active) is a Human Full Length protein, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Prothrombin, Coagulation factor II, F2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-0234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.