Learn More
Abcam Native Integrin alpha 2b + beta 3 protein (Activation site), 1MG
Supplier:  Abcam AB951301MG
Encompass_Preferred
Native Integrin alpha 2b + beta 3 protein (Activation site) is a Human Full Length protein, in the 1 to 1039 aa range, expressed in Native, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD41, GP2B, ITGAB, ITGA2B, Integrin alpha-IIb, GPalpha IIb, Platelet membrane glycoprotein IIb, GPIIb
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2255521
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.