Learn More
Abcam Native Mycoplasma Pneumoniae Protein, Full Length, Unconjugated, 200 μg
Description
- Protein Type: Full Length
- Expression System: Native
- Tested Applications: ELISA
- Form: Liquid
- Storage Buffer: PBS, pH 7 to 8
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Mycoplasma pneumoniae |
| Substrate Properties | Protein-based Substrate |
| Label or Dye | Unconjugated |
| Shipping Condition | Dry Ice |
| Form | Liquid |
| Detection Method | Colorimetric |
| Concentration | 0.25 mg/mL |
| Content And Storage | Store at -80°C. |
| Buffer | PBS, pH7 to 8 |
| Formulation | 99% PBS, pH 7 to 8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.