Learn More
Abcam Native Streptavidin Protein, DyLight 650 Conjugate, 1 mg
Description
- Protein Type: Native Streptavidin
- Expression System: Native
- Conjugation: DyLight™ 650
- Tested Applications: Flow Cyt, FRET
- Formulation: Liquid, pH 7.4
- Preservative: 0.013% Sodium azide
- Constituents: 1.42% Disodium hydrogenorthophosphate, 0.58% Sodium chloride
- Purification Technique: Size-exclusion chromatography
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | DyLight 650 |
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Biotin |
| Shipping Condition | Blue Ice |
| Detection Method | Fluorescence |
| Buffer | 1.42% disodium hydrogenorthophosphate, 0.58% sodium chloride, 0.013% sodium azide, pH7.4 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C. |
| Form | Liquid |
| Formulation | pH 7.4, 0.013% Sodium azide, 1.42% Disodium hydrogenorthophosphate, 0.58% Sodium chloride |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.