Learn More
Abcam Native Streptavidin Protein, PerCP Conjugated, Lyophilized, 1 mg
Description
- Protein Name: Streptavidin
- Source: Streptomyces avidinii
- Expression System: Native
- Applications: WB, Flow Cyt
- Purification: Size-exclusion chromatography
- Form: Lyophilized in phosphate buffer, 1.71% sucrose, 0.88% sodium chloride, 0.1% BSA, pH 7.4; contains 0.013% sodium azide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Biotin |
| Label or Dye | PerCP |
| Shipping Condition | Blue Ice |
| Form | Lyophilized |
| Detection Location | In-Blot Detection |
| Detection Method | Fluorescence |
| Buffer | phosphate buffer, 1.71% sucrose, 0.88% sodium chloride, 0.1% BSA, 0.013% sodium azide, pH7.4 |
| Content And Storage | Store at 4°C. |
| Formulation | Lyophilized from Phosphate Buffer, 1.71% sucrose, 0.88% sodium chloride, 0.1% BSA, 0.013% sodium azide, pH 7.4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.