Learn More
Hampton Research NATRIX 2 NO11 185ML
SDP
Supplier:  Hampton Research HR291711
Encompass
HR2-917-11/Natrix 2 #11 (E11), 185 ml - 0.075 M Sodium chloride, 0.002 M Calcium chloride dihydrate, 0.05 M Sodium cacodylate trihydrate pH 6.0, 30% w/v 1,6-Hexanediol, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-166-6
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.