Learn More
Description
- Contains 6 x 1.0mL vials of BK Virus NATtrol™ at concentrations of 10,000 to 1,000,000 copies/mL
- Supplied in purified protein matrix identical to the composition of true clinical specimen
Contain intact virus and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 10000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | External Run Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of Human BK virus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 6 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
