Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Coxsackievirus B5
SDP

Catalog No. 22157778 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-778 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-778 Supplier ZeptoMetrix Supplier No. NATCXVB5ST

Qualitative molecular control

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Virus Stock
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of Coxsackievirus RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.