Learn More
Description
- External run controls formulated with purified, intact and chemically modified organisms
- Non infectious and refrigerator stable
- Designed as full process extraction and amplification controls to validate molecular testing
- Supplied in purified protein matrix that mimics composition of true clinical specimen
- Evaluates performance of nucleic acid test
- 6 x 1ml vials (each control)
For verification of clinical assays, development of diagnostic tests and training of laboratory personnel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | 2°C to 8°C |
| Format | Vial |
| Product Type | External Run Control |
| Formulation | Formulated with purified, intact bacterial cells (Chlamydia trachomatis) |
| For Use With (Application) | Quality control of clinical assays and training of laboratory personnel |
| Quantity | 6 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

