Description
NATtrol™ CT/NG/TV Positive Control is formulated with purified, intact microorganisms. The microorganisms have been chemically modified to render them non-infectious and refrigerator stable. NATtrol™ CT/NG/TV Negative Control is formulated in the absence of any microorganisms. Each control pack contains 6 x 1.0 mL of NATtrol™ CT/NG/TV Positive Control or NATtrol™ CT/NG/TV Negative Control. These controls are provided in a proprietary matrix.
INTENDED USE:
- NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control are designed to evaluate the performance of nucleic acid tests for determination of the presence of C. trachomatis, N. gonorrhoeae, and T. vaginalis DNA.
- NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control enable laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Specifications
| Type | Positive Controls |
| For Use With (Application) | Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance |
RUO only.