Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Cytomegalovirus AD-169
SDP

Catalog No. 22156716 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
10000 copies/mL
100000 copies/mL
50000 copies/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-716 100000 copies/mL
22-156-886 10000 copies/mL
22-156-887 50000 copies/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-156-716 Supplier ZeptoMetrix Supplier No. NATCMV0005
Add to Cart
Add to Cart

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Each vial contains 1.0 mL of CMV NATtrol™ at target concentrations of 400 to 100,000 copies/mL
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Used to evaluate the performance of nucleic acid tests for determination of the presence of CMV DNA

Contain intact organisms and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 100000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Virus Control
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of CMV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.