Learn More
Description
- Each vial contains 1.0 mL of CMV NATtrol™ at target concentrations of 400 to 100,000 copies/mL
- Supplied in purified protein matrix identical to the composition of true clinical specimen
- Used to evaluate the performance of nucleic acid tests for determination of the presence of CMV DNA
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 10000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Virus Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of CMV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
