Learn More
Description
- Formulated with purified, intact virus particles which are chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a real clinical specimen
- Contains 6 x 1.0mL vials
- Available in different concentrations: 100, 500, 1000, 10000, 50000, 100000 copies/mL
NATEBV contains intact virus and should be run in a manner identical to that used for clinical specimens
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 50000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Virus Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of EBV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
