Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ GBS External Run Controls
GSA_VASDP

Catalog No. 21100548 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Format:
GBS Negative Result
GBS Positive Result
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
21-100-548 GBS Negative Result
21-100-558 GBS Positive Result
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-100-548 Supplier ZeptoMetrix Supplier No. NATLAC6MC
Add to Cart
Add to Cart

For determination of the presence of Group B Streptococci DNA

  • Group B Streptococci (GBS) external run controls formulated with purified, intact and chemically modified organisms
  • Non infectious and refrigerator stable
  • Designed as full process extraction and amplification controls to validate molecular testing
  • Supplied in purified protein matrix that mimics composition of true clinical specimen
  • Evaluates performance of nucleic acid test
  • 6 x 0.5ml vials (each control)

For quality control of clinical assays and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Format GBS Negative Result
Media Type Vial
Product Type External Run Control
Formulation Formulated with purified, intact bacterial cells (L.acidophilus Z048)
For Use With (Application) Determination of presence of group B Streptococci (GBS) DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.