Learn More
Description
- Gastrointestinal pathogen external run controls formulated with purified, intact and chemically modified organisms
- Non infectious and refrigerator stable
- Designed as full process extraction and amplification controls to validate molecular testing
- Supplied in stool diluent/matrix that mimics composition of true clinical specimen
- Evaluates performance of nucleic acid test
- Quantity: 0.5mL (each control)
For verification of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | 2°C to 8°C |
| Detects | Viral and bacterial Nucleic Acid |
| Form | Solution |
| Product Type | External Run Control |
| For Use With (Application) | Determination of presence of viral and bacterial Nucleic Acids, verification of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 0.5 mL |
| Strain Designations | NAP1 |
| Species | C.difficile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
