Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ GP External Run Controls
SDP

Catalog No. 21100556 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Product Type:
External Run Control
NATtrol Stool Matrix
Species:
Adenovirus Type 40
Adenovirus Type 41
C.difficile
C.jejuni
E.histolytica
Escherichia coli
Norovirus GI
Norovirus GII
Rotavirus
Salmonella typhimurium
Shigella sonnei
Strain Designations:
Clinical Isolate
DS4-868
Dugan
EDL933
ETEC; ST+, LT+
NAP1
Recombinant
TAK
Wa
Z004
Z005
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Species Strain Designations
21-100-556 External Run Control Norovirus GII Recombinant
21-100-540 External Run Control Adenovirus Type 40 Dugan
21-100-541 External Run Control Adenovirus Type 41 TAK
21-100-543 External Run Control C.jejuni Clinical Isolate
21-100-542 External Run Control C.difficile NAP1
21-100-545 External Run Control Escherichia coli EDL933
21-100-547 External Run Control E.histolytica DS4-868
21-100-546 External Run Control Escherichia coli ETEC; ST+, LT+
21-100-555 External Run Control Norovirus GI Recombinant
21-100-559 NATtrol Stool Matrix Shigella sonnei Z004
21-100-557 External Run Control Rotavirus Wa
21-100-560 NATtrol Stool Matrix Salmonella typhimurium Z005
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 21-100-556 Supplier ZeptoMetrix Supplier No. NATNOVIIGP
Add to Cart
Add to Cart

For determination of presence of viral and bacterial nucleic acids

  • Gastrointestinal pathogen external run controls formulated with purified, intact and chemically modified organisms
  • Non infectious and refrigerator stable
  • Designed as full process extraction and amplification controls to validate molecular testing
  • Supplied in stool diluent/matrix that mimics composition of true clinical specimen
  • Evaluates performance of nucleic acid test
  • Quantity: 0.5mL (each control)

For verification of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Detects Viral and bacterial Nucleic Acid
Form Solution
Media Type Vial
Product Type External Run Control
For Use With (Application) Determination of presence of viral and bacterial Nucleic Acids, verification of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 0.5 mL
Strain Designations Recombinant
Species Norovirus GII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.