Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Herpes Simplex Virus Type 1, MacIntyre Strain
SDP

Catalog No. 22156744 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
1000 copies/mL
10000 copies/mL
500 copies/mL
50000 copies/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-744 50000 copies/mL
22-156-898 10000 copies/mL
22-156-743 1000 copies/mL
22-156-742 500 copies/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-156-744 Supplier ZeptoMetrix Supplier No. NATHSV10005
Add to Cart
Add to Cart

Designed to evaluate the performance of nucleic acid tests for determination of the presence of HSV1 DNA

  • Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 1.0mL vials of target concentrations (100, 500, 1000, 10000 and 50000 copies/mL)

NATHSV1 contains intact virus and should be run in a manner identical to that used for clinical specimens

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 50000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Virus Control
For Use With (Application) Determination of presence of HSV1 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.