Learn More
ZeptoMetrix NATtrol™ Herpes Simplex Virus Type 1, MacIntyre Strain
Shop All ZeptoMetrix Corporation Products

Description
- Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Contains 1.0mL vials of target concentrations (100, 500, 1000, 10000 and 50000 copies/mL)
NATHSV1 contains intact virus and should be run in a manner identical to that used for clinical specimens
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 10000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Virus Control |
| For Use With (Application) | Determination of presence of HSV1 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.