Learn More
Description
- Formulated from highly purified infectious viruses, from cell or plasma of infected individuals
- After purification virus is chemically and enzymatically modified to alter surface proteins
- Provide completely intact viral particle that is unable to bind, penetrate or infect any host cell
Human Source Material used in the manufacture of this product has been tested and found to be negative for HBsAg, antibodies to HIV-1/2 and HCV. Handle as if capable of transmitting infectious agents; use Universal Precautions when working with this product.
Used as a control/calibrators for PCR, TMA, Branched Chain DNA and NAT assays
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 400IU/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Virus Stock |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of HBV DNA, use as controls and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
