Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Human Hepatitis C Virus
SDP

Catalog No. 22157787 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
100000IU/mL
10000IU/mL
200IU/mL
50000IU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-157-787 200IU/mL
22-157-789 10000IU/mL
22-157-790 50000IU/mL
22-157-791 100000IU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Handle as if capable of transmitting infectious agents; use Universal Precautions when working with this product.
Catalog No. 22-157-787 Supplier ZeptoMetrix Supplier No. NATHCV0001
Handle as if capable of transmitting infectious agents; use Universal Precautions when working with this product.

Ideal control for all steps of amplification assays

  • Source material is human
  • Prepared from highly purified viruses
  • Viral particles are treated chemically and enzymatically treated to alter its surface proteins and unable to bind, penetrate or infect host cell
  • Retains complete viral genome in its core

Used as controls and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays

Specifications

Certifications/Compliance ISO, 13485
Concentration 200IU/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Virus Stock
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of HCV RNA, for use as control and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.