Learn More
Description
- Non-Infectious
- Used as qualitative control
- Prepared from highly purified viruses
- Viral particles are treated chemically and enzymatically to alter their surface proteins
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | NATtrol Virus Stock |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of HHV-7 DNA, used as qualitative molecular control for nuclear assays |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
