Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Escherichia coli External Run Control
SDP

Catalog No. 22157781 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
50000cfu/mL
5000cfu/mL
For Use With (Application):
Extraction and amplification of Escherichia coli, Low, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Extraction and amplification of Escherichia coli, medium, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration For Use With (Application)
22-157-781 5000cfu/mL Extraction and amplification of Escherichia coli, Low, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
22-157-782 50000cfu/mL Extraction and amplification of Escherichia coli, medium, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-781 Supplier ZeptoMetrix Supplier No. NATECO(933)ERCL
Add to Cart
Add to Cart

Supplied in a purified serum protein matrix that mimics the composition of a true clinical specimen

  • Formulated with intact bacteria that are chemically modified to render them non-infectious and refrigerator stable
  • Calibrated against an in-house DNA standard
  • Contains 6 x 1mL vials
  • Available concentrations: 5000 or 50,000 cfu/mL

Extraction and amplification of E. coli

Specifications

Certifications/Compliance ISO, 13485
Concentration 5000cfu/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type External Run Control
Formulation Formulated with purified, intact bacterial cells
For Use With (Application) Extraction and amplification of Escherichia coli, Low, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.