Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zeptometrix NATtrol™ SARS-Related Coronavirus 2 (SARS-CoV-2) Variant B.1.351 Stock
SDP

Catalog No. 22158432 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-432 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-432 Supplier Zeptometrix Supplier No. NATSARS(COV2SA)ST
Add to Cart
Add to Cart

Designed to evaluate the performance of nucleic acid tests for determination of the presence of SARSCoV-2 Variant B.1.351 RNA.

NATtrol™ SARS-Related Coronavirus 2 (SARS-CoV-2) Variant B.1.351 Stock is formulated with purified, intact viral particles. that have been chemically modified to render them non-infectious and refrigerator stable. Each vial contains 1.0 mL of NATtrol™ SARS-CoV-2 Variant B.1.351 stock in a proprietary matrix with a target concentration.

INTENDED USE:

  • NATtrol™ SARS-CoV-2 Variant B.1.351 stock is designed to evaluate the performance of nucleic acid tests for determination of the presence of SARSCoV-2 Variant B.1.351 RNA.
  • NATtrol™ SARS-CoV2 Variant B.1.351 stock enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Concentration 1,000,000 copies/mL2
Type Stock
For Use With (Application) Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 1 mL

RUO only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.