Learn More
ZeptoMetrix NATtrol Tropical Fever Verification Panel
SDP
Catalog No. 22555240 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
9 x 0.3 mL, 3 x 1 mL
Catalog No. Quantity
22-555-240 9 x 0.3 mL, 3 x 1 mL
1 options
Catalog No. 22-555-240 Supplier ZeptoMetrix Supplier No. NATTFPBIO
Add to Cart
Add to Cart
NATtrol™ Tropical Fever Verification Panel (NATTFP-BIO) contains purified, intact bacteria and viruses that have been chemically modified to render them non-infectious and refrigerator stable. The panel members are supplied in a proprietary matrix.
NATtrol™ Tropical Fever Verification Panel (NATTFP-BIO) contains purified, intact bacteria and viruses that have been chemically modified to render them non-infectious and refrigerator stable. The panel members are supplied in a proprietary matrix.
Specifications
Type Tropical Fever Verification Panel
Quantity 9 x 0.3 mL, 3 x 1 mL
Form Liquid