Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Varicella-Zoster Virus, Strain: Ellen
SDP

Catalog No. 22156764 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
10000 copies/mL
50000 copies/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-764 10000 copies/mL
22-156-765 50000 copies/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-764 Supplier ZeptoMetrix Supplier No. NATVZV0004
Add to Cart
Add to Cart

Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable

  • Each vial contains 1.0mL of VZV NATtrol at various concentrations
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Can be used to evaluate the performance of nucleic acid tests for determination of the presence of VZV DNA

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 10000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Virus Control
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of VZV RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.