Learn More
ZeptoMetrix NATtrol™ West Nile Virus, Strain: NY 2001-6263

Shop All ZeptoMetrix Corporation Products


Description
- Each vial contains 1.0mL of WNV NATtrol at various concentrations
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Can be used to evaluate the performance of nucleic acid tests for determination of the presence of WNV RNA
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 10000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Virus Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of WNV RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.