Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Natural Chromic Gut Absorbable Sutures
SDP

Catalog No. 10000661 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
0
2-0
3-0
4-0
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-661 3-0
10-000-660 4-0
10-000-662 2-0
10-000-663 0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-661 Supplier Stoelting™ Supplier No. 50492
Add to Cart
Add to Cart

Stoelting™ Natural Chromic Gut Absorbable Sutures feature surface smoothness to facilitate knot run-down and knot strength.

  • Chromic Gut sutures are absorbable sterile surgical sutures composed of purified connective tissue (mostly collagen) derived from the serosal layer of beef (bovine) or the submucosal fibrous layer of sheep (ovine) intestines.
  • Packaged in a solution of 89% isopropanol, 10% water and 1% triethanolmine
  • Chromic Gut is treated with a chromic salt solution

The manufacturing of catgut raw material, consisting of long ribbons of highly selected serosal tissue, ensures:

  • Purity of collagen for optimal tensile strength and in vivo performance
  • Uniform treatment with a buffered chromium salt solution
  • Surface smoothness to facilitate knot run-down and knot strength.

INDICATIONS

Chromic Gut sutures are indicated for use in general soft tissue approximation and/or ligation, including use in ophthalmic surgery, but not in cardiovascular or neurological surgery.

CONTRAINDICATIONS

The use of this suture is contraindicated in patients with known sensitivities or allergies to any of its components. These sutures, being absorbable, should not be used where extended approximation of tissue is required. Chromic Gut sutures are not suitable for cardiovascular or neurological surgical procedures.

TENSILE STRENGTH

Many factors may affect the rate of tensile strength loss including type of suture, presence of inflection and tissue sites.

 

ABSORPTION

Absorption of Chromic Gut sutures occurs by an enzymatic process similar to the digestion of an animal protein by the body. Being enzymatic, the process is subject to different influencing factors (see above) and it is virtually impossible to indicate the exact designation of a consistent absorption time.

 

Specifications

For Use With (Application) Neuroscience
Length (English) 30 in.
Model 3-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.