Learn More
Creative Biomart Lepirudin
Supplier:  Creative Biomart THP0082
Encompass_Preferred
The product is identical to natural hirudin except for substitution of leucine for isoleucine at the N-terminal end of the molecule and the absence of a sulfate group on the tyrosine at position 63. It is produced via yeast cells.
Catalog No. 50-208-4133
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.