Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NAV 26

Catalog No. 469110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-911-0 10 mg
46-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-911-0 Supplier Tocris Bioscience™ Supplier No. 4691/10
This item is not returnable. View return policy

Selective Nav1.7 channel blocker

NAV 26 is a selective Nav1.7 channel blocker (IC50 = 370 nM). Exhibits >85-fold selectivity for Nav1.7 over Nav1.5 and hERG channels. Reduces nociceptive behavior in rat models of pain.

Specifications

Purity >98%
Quantity 10 mg
Product Type NAV 26
Molecular Weight (g/mol) 434.41
Inhibitors NAV 26

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.