Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAV2/RAINB1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981761
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1761 Supplier Bioss Supplier No. BS11892R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene. [supplied by OMIM].

Specifications

Antigen NAV2/RAINB1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NAV2
Gene Accession No. Q8IVL1
Gene Alias 5330421F07Rik; AU067780; BQ960892; HELAD1; Helicase APC down-regulated 1; helicase, APC down-regulated 1; KIAA1419; mKIAA3015; NAV2; Neuron navigator 2; POMFIL2; Pore membrane and/or filament-interacting-like protein 2; RAINB1; RAINB2; retinoic acid inducible gene in neuroblastoma 1; retinoic acid inducible in neuroblastoma; Retinoic acid inducible in neuroblastoma 1; STEERIN2; Steerin-2; Unc-53 homolog 2; unc53H2
Gene Symbols NAV2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NAV2/RAINB1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89797
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.