Learn More
SAPPHIRE NORTH AMERICA Naveni PD1/PD-L1 HRP
Supplier:  SAPPHIRE NORTH AMERICA NPT.PDL1.HRP.100
Encompass_Preferred
The Naveni PD1/PD-L1 is an in situ proximity ligation assay kit for chromogenic (HRP) detection of the interaction of PD1 and PD-L1. This is the first commercial assay for detecting the interaction in situ, the kit gives a high signal-to-noise ratio and has an out-standing specificity. The kit has been validated on FFPE human tissue samples. For research use only. Not for use in diagnostic procedures.
Catalog No. NC2540358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.