Learn More
SAPPHIRE NORTH AMERICA Naveni pY MET
Supplier:  SAPPHIRE NORTH AMERICA NPT.MET
Encompass_Preferred
The Naveni pTyr MET target specific assay detects phosphorylated MET with high specificity and sensitivity using our patented Naveni™ probe technology. The kit includes primary Navenibodies against phosphorylated MET and all other reagents needed to run the assay.
Catalog No. NC2540363
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.