Learn More
SAPPHIRE NORTH AMERICA NaveniBright AP
Supplier:  SAPPHIRE NORTH AMERICA NB.MR.AP.100
Encompass_Preferred
The NaveniBright AP kit is used for chromogenic visualization of protein-protein interactions, post-translational modifications like phosphorylation, or to increase the specificity when localizing single proteins. This assay is designed to be used with primary antibody pairs, generated in mouse and rabbit respectively.
Catalog No. NC2455757
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.