Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Cell GM RED
Supplier:  SAPPHIRE NORTH AMERICA NC.GM.100.RED
Encompass_Preferred
Detect proteins, protein-protein interactions, or protein modifications in cell samples. The assay is designed to be used with a goat and mouse primary antibody pair. Detection with TEX615 fluorophore is included.
Catalog No. NC2540266
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.