Learn More
SAPPHIRE NORTH AMERICA NaveniFlex control kit
Supplier:  SAPPHIRE NORTH AMERICA NF.CK
Encompass_Preferred
NaveniFlex MR Control kit is used together with NaveniFlex Cell/Tissue and Naveni Bright MR products as a control for the proximity ligation reagents, and to help the user identify positive signals.
Catalog No. NC2540355
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.