Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Tissue GM Atto647N
Supplier:  SAPPHIRE NORTH AMERICA NT.GM.100.ATTO
Encompass_Preferred
NaveniFlex Tissue GM Atto647N is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue GM Atto647N is compatible with a goat and mouse primary antibody pair and uses a Atto647N fluorophore for detection.
Catalog No. NC2540344
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.