Learn More
SAPPHIRE NORTH AMERICA NaveniFlex Tissue GR RED
Supplier:  SAPPHIRE NORTH AMERICA NT.GR.100.RED
Encompass_Preferred
NaveniFlex Tissue GR RED is designed specifically for the detection of proteins, protein interactions or modification in tissues. It improves sensitivity and removes high background seen in certain samples. NaveniFlex Tissue GR Red is compatible with a goat and rabbit primary antibody pair and uses a TEX615 fluorophore for detection.
Catalog No. NC2540270
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.