Learn More
International Enviroguard™ Navy Blue Polypropylene Coveralls with Elastic Wrists and Open Ankles
Catalog No. 17183971 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-971 3X-Large
17-183-968 Large
17-183-967 X-Large
17-183-972 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-183-971 Supplier International Enviroguard™ Supplier No. N20123XL
Add to Cart
Add to Cart
Disposable coveralls provide low-cost protection against dust, dirt, and grime.
  • Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
  • Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
  • Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
  • Elastic wrists, open ankles
  • Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
  • Stay cool, comfortable, and protected from head-to-toe
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Product Type Light-duty Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene
Color Navy
Disposable Single-use
Waist Style Standard
Ankle Style Open
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Dust
Latex Free Yes
Model N2012
No. of Pockets None