Learn More
International Enviroguard™ Navy Blue Polypropylene Coveralls with Hood and Boots
Shop All International Enviroguard Systems ProductsDescription
- Polypropylene coveralls comfortably fit over work clothes and protect against non-hazardous materials like dust, dirt, and grime
- Lightweight coverall offers a breathable alternative to traditional, bulky coverall suits
- Elastic wrists provide a secure fit to keep sleeves in place and decrease the possibility of skin exposure
- Elastic wrists, open ankles
- Disposable polypropylene coveralls are a great option for preventing cross-contamination of products, while also keeping you safe from particulates
- Stay cool, comfortable, and protected from head-to-toe
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Specifications
| Product Type | Light-duty Coveralls |
| Size | 4X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | Navy |
| Disposable | Single-use |
| Waist Style | Standard |
| Ankle Style | Attached Boots |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.