Learn More
Apexbio Technology LLC NB-598 131060-14-5 200mg
SDP
Supplier:  Apexbio Technology LLC A3645200
Encompass_Preferred
NB-598 is a selective inhibitor targeting squalene epoxidase (SE) an enzyme that catalyzes the conversion of squalene into 2 3-oxidosqualene a crucial intermediate step within the cholesterol biosynthesis pathway Through competitive inhibition of SE NB-598 interferes with the metabolic pathway associated with triglyceride synthesis involving downstream intermediates such as farnesol In experimental studies utilizing isolated murine pancreatic islets NB-598 inhibits insulin secretion under basal and glucose-stimulated conditions suggesting a functional relationship between SE activity and insulin release processes Additionally NB-598 exerts inhibitory effects on voltage-dependent calcium channels (CaV channels) pointing to a potential link between SE-mediated lipid metabolism and calcium-related cellular signaling in endocrine research This compound serves as a pharmacological tool in lipid metabolism diabetes and endocrine function studies
Catalog No. 50-000-63198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.