Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam NBD-Cl, Amine-Reactive Fluorogenic Dye, >98% Purity, Powder, 10 G

Catalog No. 502705294
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5294 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5294 Supplier Abcam Supplier No. AB14531510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amine-reactive fluorogenic dye that forms green fluorescent adducts with aliphatic amines and thiols.

  • Molecular Weight: 199.55 Da
  • Purity: >98%
  • Form: Powder
  • Reactivity: Amine-reactive, fluorogenic dye that forms green fluorescent adducts with aliphatic amines and thiols
  • Applications: Labels proteins, peptides, amino acids and other biomolecules
  • Storage: Store at ambient
  • Shipping: Ambient - can ship with ice
  • Composition: 100% 4-Chloro-7-nitrobenzo-2-oxa-1,3-diazole

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Aliphatic amines, thiols
Label or Dye NBD
Alternate Part Number 10199-89-0
Buffer 100% 4-Chloro-7-nitrobenzo-2-oxa-1,3-diazole
Content And Storage Store at ambient temperature.
Form Powder
Target Molecule Proteins, Peptides, Amino Acids, Aliphatic Amines, Thiols
Detection Method Fluorescence
Formulation 100% 4-Chloro-7-nitrobenzo-2-oxa-1,3-diazole
Quantity 10 g
For Use With (Application) Labeling proteins, peptides, amino acids and other biomolecules
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.