Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam NBD-Cl, Amine-reactive Fluorogenic Dye, Powder, 1 g

Catalog No. 502705293
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5293 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5293 Supplier Abcam Supplier No. AB1453151G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amine-reactive, fluorogenic dye that forms green fluorescent adducts with aliphatic amines and thiols to label biomolecules.

  • Molecular Weight: 199.55 Da
  • Purity: >98%
  • Form: Powder
  • Chemical Properties: Amine-reactive, fluorogenic dye that forms green fluorescent adducts with aliphatic amines and thiols
  • Applications: Labels proteins, peptides, amino acids and other biomolecules
  • Storage: Store at ambient temperature
  • Shipping: Ambient - Can ship with ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Label or Dye NBD
Buffer 100% 4-chloro-7-nitrobenzo-2-oxa-1,3-diazole
Target Molecule Proteins, Peptides, Amino Acids
Product Type Protein Labeling Reagent
Content And Storage Store at ambient temperature.
Form Powder
Substrate Properties Chemical Substrate
Substrate Aliphatic amines, thiols
Detection Method Fluorescence
Formulation 100% 4-Chloro-7-nitrobenzo-2-oxa-1,3-diazole
Quantity 1 g
For Use With (Application) Labeling proteins, peptides, amino acids and other biomolecules, Sample prep and detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.