Learn More
AdipoGen NBD-F,Chemical. CAS: 29270-56-2
SDP
Supplier:  AdipoGen CDXF0017M250
Encompass_Preferred
NBD-F, AdipoGen Life Sciences. Chemical. CAS: 29270-56-2. Formula: C6H2FN3O3. MW: 183.1. Synthetic. NBD fluoride is similar to NBD chloride except that the former is much more reactive toward amines. Both reagents are nonfluorescent until they react with primary or secondary amines to produce a fluorescent product. NBD chloride and NBD fluoride have been extensively used as derivatizing reagents for chromatography analysis of amino acids and low molecular weight amines. 29270-56-2
Catalog No. 50-117-7482
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.