Learn More
AdipoGen NBD-OMe
SDP
Supplier:  AdipoGen CDXM0520M250
Encompass_Preferred
Chemical. CAS 18333-73-8. Formula C7H5N3O4. MW 195.13. Used as intermediate or building block for synthesis of fluorescent dyes. Non-fluorescent weak fluorescent NBD variant.
Catalog No. 50-256-0457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.