Learn More
AdipoGen NBD-PZ
SDP
Supplier:  AdipoGen CDXN0014M200
Encompass_Preferred
Chemical. CAS 139332-66-4. Formula C10H11N5O3. MW 249.23. Synthetic. Fluorescent reagent for the determination of mono- and diisocyantes in air samples, and determination of lactate, hydroxybutyrate and other intermediates in body fluids. The piperazino moiety reacts readily with isocyantes to form the corresponding urea-derivatives. The derivatives can be detected by LC with UV or fluorescent-detection. Used for visualizing lysosomes in living cells also.
Catalog No. 50-256-0437
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.