Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NBEAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590713 Supplier Invitrogen™ Supplier No. PA590713
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLHWGALWRQ LASPCGAWAL RDTPIPRWKL SSAETYSRMR LKLVPNHHFD PHLEASALRD NLGEVPLTPT EEASLPLAVT KEAKVSTPPE LLQEDQLGED ELAELETPME AAELDEQREK LVLSAECQLV TVVAVVPGLL EVTTQNVYFY DGSTERVETE EGIGYDFRRP LAQLREVHLR RFNLRRSALE LFFIDQANYF LNFPCKVGTT PVSSPSQTPR PQPGPIPPHT QVRNQVYSWL LRLRPPSQGY LSSRSPQEML RASGLTQKWV QREISNFEYL MQLNTIAGRT YNDLSQYPVF P; Positive Samples: HeLa; Cellular Location: Endoplasmic reticulum.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NBEAL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NBEAL2
Gene Accession No. Q6ZNJ1
Gene Alias BDPLT4; GPS; KIAA0540; NBEAL2; neurobeachin like 2; neurobeachin-like 2; neurobeachin-like protein 2; UNQ253/PRO290
Gene Symbols NBEAL2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23218
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.