Learn More
Sigma Organic Chemistry N-Boc-trans-4-hydroxy-D-proline | 1G | 147266-92-0 | MFCD23135644
Supplier: Sigma Organic Chemistry 7620671G
N-Boc-trans-4-hydroxy-D-proline, 1G
About This Item:
Linear Formula: C10H17NO5
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.