Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBQX, Thermo Scientific™

Catalog No. p-7227391
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329260250 25 mg
AC329261000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329260250 Supplier Thermo Scientific Chemicals Supplier No. 329260250
Only null left
Add to Cart

Specifications

Chemical Name or Material NBQX
CAS 118876-58-7
Synonym 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione; 6-Nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione
Quantity 25 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C12 H8 N4 O6 S
Physical Form Powder
Color Almost-white to Yellow
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.