Learn More
ATCC NCCIT; Teratocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2073
Encompass
NCCIT is a cell line exhibiting epithelial morphology that was isolated in 1985 from the mediastinum of an Asian, adult male patient with pluripotent embryonal carcinoma. This cell line was deposited by I Damjanov and can be used in cancer research.
Catalog No. 50-238-1727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.